Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Chlorophenyl)cyclohexanone 97%

Supplier: AMBEED, INC
Warning

A315383-1G A315383-5G A315383-100MG A315383-250MG A315383-5G
103825-064EA 109 USD InStock
103825-064 103835-990 76693-676 76693-678 77306-132
4-(4-Chlorophenyl)cyclohexanone 97%
4-(4-Chlorophenyl)cyclohexanone
Precaución: for Research Use Only


Formula: C₁₂H₁₃ClO
Storage Temperature: Refrigerator
MDL Number: MFCD11850091
CAS Number: 14472-80-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty