Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorophenyl)phenylboronic acid 97%

Supplier: AMBEED, INC
Warning

A188245-1G A188245-5G A188245-250MG A188245-1G
103816-082EA 26 USD InStock
103816-082 103846-672 76998-902 77084-400
4-(4-Fluorophenyl)phenylboronic acid 97%
4-(4-Fluorophenyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₀BFO₂
MW: 216.018 g/mol
Boiling Pt: 382.3 °C (760 mm Hg)
Melting Pt: 212…250 °C
Flash Pt: 185 °C
Storage Temperature: Freezer
MDL Number: MFCD08544387
CAS Number: 140369-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty