Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Fluorophenyl)piperidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A236170-1G A236170-5G A236170-5G A236170-250MG A236170-1G
103811-072EA 36 USD InStock
103811-072 103850-842 77089-884 77113-120 77304-032
4-(4-Fluorophenyl)piperidine hydrochloride 97%
4-(4-Fluorophenyl)piperidine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₁₅ClFN
Storage Temperature: Ambient
MDL Number: MFCD01632528
CAS Number: 6716-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty