Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4-Methoxyphenyl)-1-butanol 98%

Supplier: AMBEED, INC
Warning

A178334-1G A178334-5G A178334-1G A178334-100G
103807-322EA 15 USD InStock
103807-322 103845-662 77301-554 77671-524
4-(4-Methoxyphenyl)-1-butanol 98%
4-(4-Methoxyphenyl)-1-butanol
Precaución: for Research Use Only


Formula: C₁₁H₁₆O₂
Boiling Pt: 304.6 °C (760 mm Hg)
Melting Pt: 3…4 °C
Flash Pt: 130.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00002972
CAS Number: 52244-70-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty