Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Morpholino-5-pyridineboronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A162728-1G A162728-5G A162728-250MG A162728-1G
103805-654EA 16 USD InStock
103805-654 103843-904 77163-246 77319-976
2-Morpholino-5-pyridineboronic acid pinacol ester 97%
2-Morpholino-5-pyridineboronic acid pinacol ester


Formula: C₁₅H₂₃BN₂O₃
Boiling Pt: 440.9 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04039874
CAS Number: 485799-04-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty