Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)benzonitrile 97%

Supplier: AMBEED, INC
Warning

A111198-1G A111198-100G A111198-10G A111198-25G A111198-1G A111198-5G
103797-798EA 6 USD InStock
103797-798 103823-610 77245-706 77245-708 77245-710 77310-414 77310-416
4-(Aminomethyl)benzonitrile 97%
4-(Aminomethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₈N₂
MW: 132.1625 g/mol
Boiling Pt: 275.4 °C (760 mm Hg)
Melting Pt: 274…279 °C
Flash Pt: 120.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00025578
CAS Number: 10406-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty