Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Benzyloxy)aniline hydrochloride 99%

Supplier: AMBEED, INC
Warning

A118850-100G A118850-10G A118850-25G A118850-500G
76971-376EA 55 USD InStock
76971-376 76971-378 76971-380 77880-963
4-(Benzyloxy)aniline hydrochloride 99%
4-(Benzyloxy)aniline hydrochloride
Precaución: for Research Use Only


Formula: C₁₃H₁₄ClNO
MW: 235.711 g/mol
Melting Pt: 228 °C
Flash Pt: 180.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00012995
CAS Number: 51388-20-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty