Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Bis(4-fluorophenyl)methylene)piperidine 95%

Supplier: AMBEED, INC
Warning

A187105-1G A187105-5G A187105-100MG A187105-250MG
103815-992EA 63 USD InStock
103815-992 103846-584 76985-962 76985-964
4-(Bis(4-fluorophenyl)methylene)piperidine 95%
4-(Bis(4-fluorophenyl)methylene)piperidine
Precaución: for Research Use Only


Formula: C₁₈H₁₇F₂N
Boiling Pt: 404.4±40.0 °C (760 mm Hg)
Flash Pt: 198.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13192725
CAS Number: 58113-36-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty