Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Butylamino)benzoic acid 97%

Supplier: AMBEED, INC
Warning

A132317-1G A132317-250MG A132317-1G A132317-25G A132317-5G A132317-100G
103801-716EA 33 USD InStock
103801-716 77285-366 77314-292 77314-294 77314-296 77747-944
4-(Butylamino)benzoic acid 97%
4-(Butylamino)benzoic acid
Precaución: for Research Use Only


Formula: C₁₁H₁₅NO₂
MW: 193.242 g/mol
Melting Pt: 149…151 °C
Flash Pt: 167.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00002536
CAS Number: 4740-24-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty