Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Terephthalaldehyde mono(diethyl acetal) 97%

Supplier: AMBEED, INC
Warning

A100588-1G A100588-5G A100588-10G A100588-25G A100588-1G
103795-526EA 10 USD InStock
103795-526 103821-214 76717-490 76717-492 77296-768
Terephthalaldehyde mono(diethyl acetal) 97%
Terephthalaldehyde mono(diethyl acetal)
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₃
Boiling Pt: 290.3 °C (760 mm Hg)
Flash Pt: 120.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00010217
CAS Number: 81172-89-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty