Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Dimethylamino)butanoic acid hydrochloride 97%

Supplier: AMBEED, INC
Warning

A122464-100G A122464-10G A122464-25G A122464-500G A122464-5G
77149-268EA 73 USD InStock
77149-268 77149-270 77149-272 77149-274 77312-362
4-(Dimethylamino)butanoic acid hydrochloride 97%
4-(Dimethylamino)butanoic acid hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₄ClNO₂
MW: 167.64 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00012613
CAS Number: 69954-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty