Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Ethylamino)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A125325-1G A125325-100MG A125325-250MG A125325-1G A125325-5G
103800-668EA 210 USD InStock
103800-668 76822-666 76822-668 77298-042 77298-044
4-(Ethylamino)benzaldehyde 98%
4-(Ethylamino)benzaldehyde
Precaución: for Research Use Only


Formula: C₉H₁₁NO
Storage Temperature: Refrigerator
MDL Number: MFCD18447689
CAS Number: 79865-89-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty