Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hydroxymethyl)benzonitrile 98%

Supplier: AMBEED, INC
Warning

A295046-1G A295046-1G A295046-5G A295046-10G A295046-25G
103814-040EA 13 USD InStock
103814-040 103835-176 76837-822 76837-824 77289-854 77305-666
4-(Hydroxymethyl)benzonitrile 98%
4-(Hydroxymethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 297.4 °C (760 mm Hg)
Melting Pt: 39…43 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00870633
CAS Number: 874-89-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty