Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Hydroxymethyl)thiazole 97%

Supplier: AMBEED, INC
Warning

A193184-1G A193184-5G A193184-10G A193184-1G A193184-25G A193184-5G A193184-250MG
103816-508EA 63 USD InStock
103816-508 103847-122 77287-520 77302-328 77302-330 77302-332 77748-228
4-(Hydroxymethyl)thiazole 97%
4-(Hydroxymethyl)thiazole
Precaución: for Research Use Only


Formula: C₄H₅NOS
Boiling Pt: 248.5 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115728
CAS Number: 7036-04-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty