Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methoxymethoxy)benzoic acid 98%

Supplier: AMBEED, INC
Warning

A160862-1G A160862-5G A160862-10G A160862-250MG A160862-25G
103805-426EA 15 USD InStock
103805-426 103843-660 77138-156 77138-158 77138-160
4-(Methoxymethoxy)benzoic acid 98%
4-(Methoxymethoxy)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₄
Storage Temperature: Ambient
MDL Number: MFCD21101751
CAS Number: 25458-44-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty