Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methoxymethyl)aniline 97%

Supplier: AMBEED, INC
Warning

A202742-1G A202742-5G A202742-10G A202742-1G A202742-25G A202742-250MG
103817-410EA 47 USD InStock
103817-410 103848-054 76999-406 77302-818 77302-820 77881-478
4-(Methoxymethyl)aniline 97%
4-(Methoxymethyl)aniline
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MW: 137.179 g/mol
Boiling Pt: 224.9 °C (760 mm Hg)
Melting Pt: 38…39 °C
Flash Pt: 93.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06804487
CAS Number: 80936-82-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty