Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A175498-1G A175498-5G A175498-100MG A175498-10G A175498-250MG
103807-046EA 36 USD InStock
103807-046 103845-354 77011-738 77011-740 77011-742
4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde 98%
4-(Methylamino)-2-(methylthio)pyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₇H₉N₃OS
MW: 183.23 g/mol
Boiling Pt: 377.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04115117
CAS Number: 185040-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty