Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Methylsulfonyl)benzaldehyde 97%

Supplier: AMBEED, INC
Warning

A160410-500G A160410-25G A160410-100G
103808-884EA 54 USD InStock
103808-884 77247-648 77300-462 77746-095
4-(Methylsulfonyl)benzaldehyde 97%
4-(Methylsulfonyl)benzaldehyde
Precaución: for Research Use Only


Formula: C₈H₈O₃S
Boiling Pt: 378.3 °C (760 mm Hg)
Melting Pt: 155…161 °C
Flash Pt: 242.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00216485
CAS Number: 5398-77-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty