Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Pyridin-2-yl)butanoic acid 95%

Supplier: AMBEED, INC
Warning

A123194-1G A123194-10G A123194-250MG A123194-1G A123194-5G
103800-360EA 57 USD InStock
103800-360 77026-588 77026-590 77312-478 77312-480
4-(Pyridin-2-yl)butanoic acid 95%
4-(Pyridin-2-yl)butanoic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
Boiling Pt: 303.7 °C (760 mm Hg)
Flash Pt: 137.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234552
CAS Number: 102879-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty