Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(trans-4-Butylcyclohexyl)phenol 97%

Supplier: AMBEED, INC
Warning

A206462-25G A206462-5G A206462-100G A206462-10G
103848-384EA 6 USD InStock
103848-384 77086-644 77086-646 77275-002 77275-004
4-(trans-4-Butylcyclohexyl)phenol 97%
4-(trans-4-Butylcyclohexyl)phenol
Precaución: for Research Use Only


Formula: C₁₆H₂₄O
MW: 232.37 g/mol
Boiling Pt: 354.8 °C (760 mm Hg)
Flash Pt: 186.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00673752
CAS Number: 88581-00-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty