Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)-1H-pyrazole 97%

Supplier: AMBEED, INC
Warning

A674313-5G A674313-100G A674313-250MG A674313-1G A674313-25G
103840-512EA 112 USD InStock
103840-512 76844-706 77005-812 77005-814 77308-102 77308-104
4-(Trifluoromethyl)-1H-pyrazole 97%
4-(Trifluoromethyl)-1H-pyrazole
Precaución: for Research Use Only


Formula: C₄H₃F₃N₂
MW: 136.077 g/mol
Boiling Pt: 177.7±35.0 °C (760 mm Hg)
Melting Pt: 73…78 °C
Flash Pt: 61.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11226572
CAS Number: 52222-73-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty