Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Trifluoromethyl)hydrocinnamic acid 98%

Supplier: AMBEED, INC
Warning

A114179-1G A114179-5G A114179-10G A114179-1G A114179-5G A114179-100G
103798-358EA 6 USD InStock
103798-358 103824-198 76797-006 77297-290 77311-018 77878-859
4-(Trifluoromethyl)hydrocinnamic acid 98%
4-(Trifluoromethyl)hydrocinnamic acid
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O₂
MW: 218.17 g/mol
Boiling Pt: 279 °C (760 mm Hg)
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00674032
CAS Number: 53473-36-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty