Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(α,α,α-Trifluoro-4-tolyl)acetic acid 98%

Supplier: AMBEED, INC
Warning

A124150-1G A124150-100G A124150-10G A124150-25G A124150-500G A124150-1G A124150-5G
103800-490EA 9 USD InStock
103800-490 76688-252 76688-254 76688-256 76688-258 77297-972 77297-974
(α,α,α-Trifluoro-4-tolyl)acetic acid 98%
(α,α,α-Trifluoro-4-tolyl)acetic acid
Precaución: for Research Use Only


Formula: (C₆H₄(CF₃))CH₂COOH
MW: 204.15 g/mol
Boiling Pt: 263.1 °C (760 mm Hg)
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004352
CAS Number: 32857-62-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty