Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Naphthyl boronic acid pinacol ester 97%

Supplier: AMBEED, INC
Warning

A101789-1G A101789-5G A101789-100G A101789-250MG A101789-25G A101789-1G A101789-5G
103796-136EA 6 USD InStock
103796-136 103821-824 76717-574 76717-576 76717-578 77308-718 77308-720
2-Naphthyl boronic acid pinacol ester 97%
2-Naphthyl boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₆H₁₉BO₂
MW: 254.14 g/mol
Boiling Pt: 378.7 °C (760 mm Hg)
Flash Pt: 182.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05663868
CAS Number: 256652-04-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty