Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydropyran-4-boronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A106833-1G A106833-100MG A106833-250MG A106833-5G A106833-1G
103797-086EA 65 USD InStock
103797-086 76809-672 76809-674 76809-676 77309-698
Tetrahydropyran-4-boronic acid pinacol ester 95%
Tetrahydropyran-4-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₁H₂₁BO₃
Boiling Pt: 251.8±33.0 °C (760 mm Hg)
Flash Pt: 106.1 °C
Storage Temperature: Freezer
MDL Number: MFCD11506065
CAS Number: 1131912-76-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty