Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dibromo-2,2'-bipyridyl 98%

Supplier: AMBEED, INC
Warning

A359722-1G A359722-1G A359722-25G A359722-5G A359722-100G A359722-100MG A359722-10G
103826-232EA 16 USD InStock
103826-232 103837-240 76840-420 76840-422 76840-424 77290-752 77290-754 77290-756 77290-758
4,4'-Dibromo-2,2'-bipyridyl 98%
4,4'-Dibromo-2,2'-bipyridyl
Precaución: for Research Use Only


Formula: C₁₀H₆Br₂N₂
Boiling Pt: 362.9 °C (760 mm Hg)
Melting Pt: 138 °C
Flash Pt: 173.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00667763
CAS Number: 18511-71-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty