Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3,5-Tris(4-bromophenyl)benzene 97%

Supplier: AMBEED, INC
Warning

A160503-1G A160503-5G A160503-100G A160503-10G A160503-250MG A160503-1G A160503-25G A160503-5G
103805-386EA 11 USD InStock
103805-386 103843-618 76871-978 76871-980 76871-982 77319-558 77319-560 77319-562
1,3,5-Tris(4-bromophenyl)benzene 97%
1,3,5-Tris(4-bromophenyl)benzene
Precaución: for Research Use Only


Formula: C₂₄H₁₅Br₃
Boiling Pt: 574.9 °C (760 mm Hg)
Melting Pt: 261…265 °C
Storage Temperature: Ambient
MDL Number: MFCD00362911
CAS Number: 7511-49-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty