Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dibromotriphenylamine 98%

Supplier: AMBEED, INC
Warning

A118842-1G A118842-100G A118842-10G A118842-25G A118842-1G A118842-5G
103799-114EA 6 USD InStock
103799-114 77284-698 77284-700 77284-702 77311-786 77311-788
4,4'-Dibromotriphenylamine 98%
4,4'-Dibromotriphenylamine
Precaución: for Research Use Only


Formula: C₁₈H₁₃Br₂N
Melting Pt: 80…84 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00060104
CAS Number: 81090-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty