Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Difluorocyclohexylamine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A377431-1G A377431-25G A377431-5G A377431-10G A377431-250MG
103826-608EA 10 USD InStock
103826-608 103837-638 76840-946 76840-948 76840-950 77290-942 77290-944
4,4-Difluorocyclohexylamine hydrochloride 98%
4,4-Difluorocyclohexylamine hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂ClF₂N
Melting Pt: 295 °C
Flash Pt: 67.4 °C
Storage Temperature: Ambient
MDL Number: MFCD06246881
CAS Number: 675112-70-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty