Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dimethoxy-2,2'-bipyridyl 98%

Supplier: AMBEED, INC
Warning

A346917-1G A346917-1G A346917-5G A346917-100G A346917-10G A346917-250MG
103825-942EA 14 USD InStock
103825-942 103836-930 76839-996 76839-998 77003-210 77003-212 77003-214
4,4'-Dimethoxy-2,2'-bipyridyl 98%
4,4'-Dimethoxy-2,2'-bipyridyl
Precaución: for Research Use Only


Formula: C₁₂H₁₂N₂O₂
Boiling Pt: 347.6 °C (760 mm Hg)
Melting Pt: 168…171 °C
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00233880
CAS Number: 17217-57-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty