Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4'-Dimethoxybenzil 98%

Supplier: AMBEED, INC
Warning

A300247-5G A300247-100G A300247-10G A300247-25G A300247-5G A300247-500G
103835-416EA 7 USD InStock
103835-416 77032-476 77032-478 77305-796 77305-798 77872-398
4,4'-Dimethoxybenzil 98%
4,4'-Dimethoxybenzil
Precaución: for Research Use Only


Formula: C₁₆H₁₄O₄
MW: 270.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008405
CAS Number: 1226-42-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty