Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dimethyldihydrofuran-2,3-dione 95%

Supplier: AMBEED, INC
Warning

A123111-1G A123111-250MG A123111-1G A123111-5G
103800-342EA 19 USD InStock
103800-342 76797-506 77297-922 77297-924
4,4-Dimethyldihydrofuran-2,3-dione 95%
4,4-Dimethyldihydrofuran-2,3-dione
Precaución: For research use only.


Formula: C₆H₈O₃
Boiling Pt: 260.5 °C (760 mm Hg)
Melting Pt: 67…69 °C
Flash Pt: 122.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012331
CAS Number: 13031-04-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty