Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethylglutarimide 98%

Supplier: AMBEED, INC
Warning

A112146-1G A112146-10G A112146-1G A112146-5G A112146-25G
103797-962EA 7 USD InStock
103797-962 103823-796 77270-972 77284-212 77310-604 77747-823
3,3-Dimethylglutarimide 98%
3,3-Dimethylglutarimide
Precaución: for Research Use Only


Formula: C₇H₁₁NO₂
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00006671
CAS Number: 1123-40-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty