Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydro-1H-benzoimidazole 98%

Supplier: AMBEED, INC
Warning

A122859-1G A122859-100MG A122859-250MG A122859-25G A122859-1G A122859-10G
103800-294EA 18 USD InStock
103800-294 77284-930 77284-932 77284-934 77312-416 77747-888
4,5,6,7-Tetrahydro-1H-benzoimidazole 98%
4,5,6,7-Tetrahydro-1H-benzoimidazole
Precaución: for Research Use Only


Formula: C₇H₁₀N₂
Boiling Pt: 335 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00829762
CAS Number: 3752-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty