Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 97%

Supplier: AMBEED, INC
Warning

A216443-1G A216443-5G A216443-100G A216443-1G A216443-25G A216443-5G
103818-558EA 6 USD InStock
103818-558 103849-278 76770-698 77087-860 77087-862 77087-864
2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole 97%
2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole
Precaución: for Research Use Only


Formula: C₇H₁₁N₃S
Boiling Pt: 359 °C (760 mm Hg)
Melting Pt: 217…225 °C
Flash Pt: 170.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07771983
CAS Number: 104617-49-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty