Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5,6,7-Tetrahydrothieno[2,3-c]pyridine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A117517-1G A117517-100MG A117517-10G A117517-250MG A117517-1G A117517-5G
103798-906EA 56 USD InStock
103798-906 103824-794 77284-620 77284-622 77284-624 77297-548 77297-550
4,5,6,7-Tetrahydrothieno[2,3-c]pyridine hydrochloride 98%
4,5,6,7-Tetrahydrothieno[2,3-c]pyridine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₀ClNS
Flash Pt: 122.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08448167
CAS Number: 28783-38-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty