Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,5-Dichloropyrimidine 97%

Supplier: AMBEED, INC
Warning

A253527-1G A253527-5G A253527-1G A253527-5G A253527-100MG A253527-250MG
103812-096EA 49 USD InStock
103812-096 103851-946 77091-380 77091-382 77113-512 77113-514
4,5-Dichloropyrimidine 97%
4,5-Dichloropyrimidine
Precaución: for Research Use Only


Formula: C₄H₂Cl₂N₂
Boiling Pt: 219.7 °C (760 mm Hg)
Flash Pt: 107.572 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09743880
CAS Number: 6554-61-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty