Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Diamino-5-nitropyrimidine 95%

Supplier: AMBEED, INC
Warning

A165810-1G A165810-5G A165810-1G A165810-5G
103806-002EA 15 USD InStock
103806-002 103844-256 77320-480 77320-482
4,6-Diamino-5-nitropyrimidine 95%
4,6-Diamino-5-nitropyrimidine
Precaución: for Research Use Only


Formula: C₄H₅N₅O₂
Boiling Pt: 461.4 °C (760 mm Hg)
Flash Pt: 232.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00023268
CAS Number: 2164-84-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty