Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-2,3-dihydro-1H-pyrrolo[3,4-c]pyridin-1-one 95%

Supplier: AMBEED, INC
Warning

A198498-25MG A198498-1G A198498-100MG A198498-250MG A198498-50MG A198498-1G A198498-25MG
103792-368EA 25 USD InStock
103792-368 103817-026 103847-652 76873-306 76873-308 76873-310 77085-682 77085-684
4,6-Dichloro-2,3-dihydro-1H-pyrrolo[3,4-c]pyridin-1-one 95%
4,6-Dichloro-2,3-dihydro-1H-pyrrolo[3,4-c]pyridin-1-one
Precaución: for Research Use Only


Formula: C₇H₄Cl₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD16658728
CAS Number: 1201676-03-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty