Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde 97%

Supplier: AMBEED, INC
Warning

A149907-1G A149907-5G A149907-10G A149907-250MG A149907-1G A149907-25G A149907-5G
103804-100EA 14 USD InStock
103804-100 103842-226 76973-054 76973-056 77317-742 77317-744 77317-746
4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde 97%
4,6-Dichloro-2-methylpyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₆H₄Cl₂N₂O
Storage Temperature: Freezer
MDL Number: MFCD09909782
CAS Number: 14160-91-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty