Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloroisatin 98%

Supplier: AMBEED, INC
Warning

A194513-1G A194513-5G A194513-1G A194513-5G A194513-10G
103816-660EA 16 USD InStock
103816-660 103847-270 77085-186 77085-188 77671-724
4,6-Dichloroisatin 98%
4,6-Dichloroisatin
Precaución: for Research Use Only


Formula: C₈H₃Cl₂NO₂
Melting Pt: 260.5…261 °C
Storage Temperature: Ambient
MDL Number: MFCD01971146
CAS Number: 18711-15-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty