Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dihydroxyisophthalic acid 97%

Supplier: AMBEED, INC
Warning

A170184-1G A170184-5G A170184-10G A170184-250MG A170184-1G A170184-25G A170184-5G
103806-456EA 8 USD InStock
103806-456 103844-722 76824-466 76824-468 77081-828 77081-830 77081-832
4,6-Dihydroxyisophthalic acid 97%
4,6-Dihydroxyisophthalic acid
Precaución: for Research Use Only


Formula: C₈H₆O₆
Boiling Pt: 551.0±35.0 °C (760 mm Hg)
Melting Pt: 312 °C
Flash Pt: 301.1 °C
Storage Temperature: Ambient
MDL Number: MFCD08693472
CAS Number: 19829-74-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty