Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dimethylnicotinic acid 98%

Supplier: AMBEED, INC
Warning

A140498-1G A140498-10G A140498-250MG A140498-1G A140498-5G
103802-878EA 62 USD InStock
103802-878 77285-714 77285-716 77315-990 77315-992
4,6-Dimethylnicotinic acid 98%
4,6-Dimethylnicotinic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
Boiling Pt: 310 °C (760 mm Hg)
Melting Pt: 193…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00234424
CAS Number: 22047-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty