Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,7-Dimethyl-1,10-phenanthroline 98%

Supplier: AMBEED, INC
Warning

A158709-1G A158709-5G A158709-250MG A158709-25G A158709-5G
103805-162EA 50 USD InStock
103805-162 103843-376 77286-368 77300-360 77300-362
4,7-Dimethyl-1,10-phenanthroline 98%
4,7-Dimethyl-1,10-phenanthroline
Precaución: for Research Use Only


Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Boiling Pt: 397.5 °C (760 mm Hg)
Melting Pt: 193…195 °C
Flash Pt: 176.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00004979
CAS Number: 3248-05-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty