Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetyl-6-tert-butyl-1,1-dimethylindan 98%

Supplier: AMBEED, INC
Warning

A381702-5G A381702-1G A381702-100G
103837-738EA 28 USD InStock
103837-738 76841-056 77033-430 77668-764
4-Acetyl-6-tert-butyl-1,1-dimethylindan 98%
4-Acetyl-6-tert-butyl-1,1-dimethylindan
Precaución: for Research Use Only


Formula: C₁₇H₂₄O
Boiling Pt: 309.1 °C (760 mm Hg)
Melting Pt: 68…70 °C at N/A mmHg
Flash Pt: 125.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00046324
CAS Number: 13171-00-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty