Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Acetylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A205870-25G A205870-5G A205870-100G A205870-10G A205870-500G
103848-338EA 14 USD InStock
103848-338 77086-568 77086-570 77260-912 77260-914 77260-916
4-Acetylbenzoic acid 97%
4-Acetylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₈O₃
MW: 164.16 g/mol
Boiling Pt: 339.9 °C (760 mm Hg)
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002561
CAS Number: 586-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty