Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-1-isopropylpiperidine 97%

Supplier: AMBEED, INC
Danger

A457549-5G A457549-1G A457549-25G A457549-5G
103827-876EA 26 USD InStock
103827-876 103838-952 76842-816 76842-818 76842-820
4-Amino-1-isopropylpiperidine 97%
4-Amino-1-isopropylpiperidine
Precaución: for Research Use Only


Formula: C₈H₁₈N₂
Boiling Pt: 273.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD03411606
CAS Number: 127285-08-9
UN: 2920
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty