Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%

Supplier: AMBEED, INC
Warning

A160341-1G A160341-5G A160341-10G A160341-250MG A160341-1G A160341-25G A160341-5G
103805-368EA 8 USD InStock
103805-368 103843-596 77286-420 77286-422 77319-522 77319-524 77319-526
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde 98%
4-Amino-2-(methylthio)pyrimidine-5-carbaldehyde
Precaución: for Research Use Only


Formula: C₆H₇N₃OS
Boiling Pt: 379.7 °C (760 mm Hg)
Flash Pt: 183.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08669438
CAS Number: 770-31-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty