Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-(trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Warning

A115019-1G A115019-5G A115019-100G A115019-10G A115019-250MG A115019-25G A115019-1G A115019-5G
103798-516EA 9 USD InStock
103798-516 103824-360 77257-872 77257-874 77257-876 77257-878 77311-176 77311-178
4-Amino-2-(trifluoromethyl)pyridine 98%
4-Amino-2-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₅F₃N₂
Boiling Pt: 230.7 °C (760 mm Hg)
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD09056877
CAS Number: 147149-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty