Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3,5-dimethylbenzoic acid 98%

Supplier: AMBEED, INC
Warning

A170827-1G A170827-5G A170827-100MG A170827-250MG A170827-5G
103806-548EA 28 USD InStock
103806-548 103844-824 76974-002 76974-004 77081-946
4-Amino-3,5-dimethylbenzoic acid 98%
4-Amino-3,5-dimethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 342.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00047840
CAS Number: 4919-40-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty